accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177-10-6

OO

CAS RN 177-10-6

IUPAC Name: 1,4-dioxaspiro[4.5]decane
Synonyms: 1,4-dioxaspiro(4.5)decane
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: GZGPRZYZKBQPBQ-UHFFFAOYSA-N
SMILES: C1COC2(CCCCC2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results