Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Pentamethylene-1,3-dioxolane, 99%, Thermo Scientific™

Catalog No. AC223350250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC223350250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC223350250 Supplier Thermo Scientific Chemicals Supplier No. 223350250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177-10-6
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
InChI Key GZGPRZYZKBQPBQ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decane, cyclohexanone ethylene ketal, 2,2-pentamethylene-1,3-dioxolane, cyclohexanone ethylene acetal, ethylenedioxy cyclohexane, spiro cyclohexane-1,2'-1,3 dioxolane, cyclohexanone ethlyene ketal, 5-19-01-00222 beilstein handbook reference
PubChem CID 9093
IUPAC Name 1,4-dioxaspiro[4.5]decane
SMILES C1CCC2(CC1)OCCO2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.02g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4575 to 1.4595 (20°C, 589nm)
Quantity 25 g
Beilstein 19, III, 128
Formula Weight 142.2
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2, 2-Pentamethylene-1, 3-dioxolane, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.