accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3ClClClClClCl

CAS RN 1770-80-5

IUPAC Name: 2,3-dibutyl 1,4,5,6,7,7-hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Synonyms: 2,3-dibutyl 1,4,5,6,7,7-hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Molecular Weight (g/mol): 501.04
Molecular Formula: C17H20Cl6O4
InChi Key: UJAHPBDUQZFDLA-UHFFFAOYNA-N
SMILES: CCCCOC(=O)C1C(C(=O)OCCCC)C2(Cl)C(Cl)=C(Cl)C1(Cl)C2(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results