accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177202-83-4

IUPAC Name: 2-(pyridin-3-yl)aniline
Synonyms: 2-(pyridin-3-yl)aniline
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: MTYHJHATMPCNLW-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results