accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 17739-45-6

IUPAC Name: 2-(2-bromoethoxy)oxane
Synonyms: 2-(2-bromoethoxy)oxane
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: GCUOLJOTJRUDIZ-UHFFFAOYNA-N
SMILES: BrCCOC1CCCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results