Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromoethoxy)tetrahydro-2H-pyran (stabilized with K2CO3) 95.0+%, TCI America™
SDP

Catalog No. B28505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28505G 5 g
B285025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28505G Supplier TCI America Supplier No. B28505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17739-45-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
InChI Key GCUOLJOTJRUDIZ-UHFFFAOYSA-N
Synonym 2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran
PubChem CID 86621
IUPAC Name 2-(2-bromoethoxy)oxane
SMILES C1CCOC(C1)OCCBr

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 209.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromoethoxy)tetrahydro-2H-pyran (stabilized with K2CO3)
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.