accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1774-47-6

IUPAC Name: trimethyl(oxo)-λ⁶-sulfanylium iodide
Synonyms: trimethylsulfoxonium iodide
Molecular Weight (g/mol): 220.07
Molecular Formula: C3H9IOS
InChi Key: BPLKQGGAXWRFOE-UHFFFAOYSA-M
SMILES: [I-].C[S+](C)(C)=O
Percent Purity 
  • (3)
  • (3)
  • (5)
Quantity 
  • (4)
  • (3)
  • (4)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (5)
  • (4)
  • (3)
  • (4)
  • (5)