accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17747-43-2

IUPAC Name: pyridin-3-yl acetate
Synonyms: pyridin-3-yl acetate
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: QZDWODWEESGPLC-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results