Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetoxypyridine 98.0+%, TCI America™
SDP

Catalog No. A121025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A121025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A121025G Supplier TCI America Supplier No. A121025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17747-43-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006377
InChI Key QZDWODWEESGPLC-UHFFFAOYSA-N
Synonym 3-Pyridyl Acetate
PubChem CID 87289
IUPAC Name pyridin-3-yl acetate
SMILES CC(=O)OC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Acetoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.