accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177480-81-8

IUPAC Name: 4-bromo-2-chloro-5-fluorobenzoic acid
Synonyms: 4-bromo-2-chloro-5-fluorobenzoic acid
Molecular Weight (g/mol): 253.45
Molecular Formula: C7H3BrClFO2
InChi Key: TZFKKLGCVKORLS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(Br)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results