Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chloro-5-fluorobenzoic acid, 98%, Thermo Scientific™

Catalog No. AAH6396514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6396514 25 g
AAH6396503 1 g
AAH6396506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6396514 Supplier Thermo Scientific Chemicals Supplier No. H6396514

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177480-81-8
Molecular Formula C7H3BrClFO2
Molecular Weight (g/mol) 253.45
MDL Number MFCD00236589
InChI Key TZFKKLGCVKORLS-UHFFFAOYSA-N
Synonym 4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
PubChem CID 2736225
IUPAC Name 4-bromo-2-chloro-5-fluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(Br)C=C1Cl

Specifications

Color White
Quantity 25 g
Formula Weight 253.45
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-chloro-5-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.