accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 1775-27-5

IUPAC Name: 2-bromo-2,3-dihydro-1H-inden-1-one
Synonyms: 2-bromo-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: UXVCSPSWUNMPMT-UHFFFAOYNA-N
SMILES: BrC1CC2=CC=CC=C2C1=O
Boiling Point 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results