Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-indanone, tech. 85%, Thermo Scientific™

Catalog No. AAL1245303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1245303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1245303 Supplier Thermo Scientific Chemicals Supplier No. L1245303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1775-27-5
Melting Point 37°C to 39°C
Boiling Point 110°C to 112°C (0.4 mmHg)
Molecular Formula C9H7BrO
Quantity 1 g
MDL Number MFCD00041030
Beilstein 637789
Synonym 2-bromo-1-indanone, 2-bromoindan-1-one, 2-bromoindanone, 2-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 2-bromo-2,3-dihydro, bromoindanone, 2-bromo indanone, pubchem8852, 2-bromo-indan-1-one, acmc-20a5au
InChI Key UXVCSPSWUNMPMT-UHFFFAOYSA-N
SMILES C1C(C(=O)C2=CC=CC=C21)Br
IUPAC Name 2-bromo-2,3-dihydroinden-1-one
Molecular Weight (g/mol) 211.058
PubChem CID 137203
Formula Weight 211.06
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Bromo-1-indanone
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.