accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177582-21-7

SOOOOHONH(R)CH3

CAS RN 177582-21-7

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(4-methoxyphenyl)diphenylmethyl]sulfanyl}propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-{[(4-methoxyphenyl)diphenylmethyl]sulfanyl}propanoic acid
Molecular Weight (g/mol): 615.74
Molecular Formula: C38H33NO5S
InChi Key: LOBUWFUSGOYXQX-DHUJRADRSA-N
SMILES: COC1=CC=C(C=C1)C(SC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)