accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1776-37-0

IUPAC Name: 5-methyl-1H-indazole
Synonyms: 5-methyl-1H-indazole
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: DCUNRLLJHAWKRZ-UHFFFAOYSA-N
SMILES: CC1=CC=C2NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results