CAS RN 177609-12-0
CAS RN 177609-12-0
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid
Molecular Weight (g/mol):
395.41
Molecular Formula:
C22H21NO6
InChi Key:
FBNFRRNBFASDKS-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Asp OAll -OH, Advanced ChemTech
CAS: 177609-12-0 Molecular Formula: C22H21NO6 Molecular Weight (g/mol): 395.41 InChI Key: FBNFRRNBFASDKS-UHFFFAOYNA-N IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid SMILES: OC(=O)C(CC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12