accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17763-80-3

IUPAC Name: 4-nitrophenyl trifluoromethanesulfonate
Synonyms: 4-nitrophenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 271.17
Molecular Formula: C7H4F3NO5S
InChi Key: NDTIXHNCNLKURN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)