accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177743-06-5

IUPAC Name: 3-chloro-2-ethoxypyridine
Synonyms: 3-chloro-2-ethoxypyridine
Molecular Weight (g/mol): 157.6
Molecular Formula: C7H8ClNO
InChi Key: MTTKGSCSPHCOOF-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results