accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177966-61-9

SOOOHONH(R)

CAS RN 177966-61-9

IUPAC Name: (2R)-3-(1-benzothiophen-3-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2R)-3-(1-benzothiophen-3-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 443.52
Molecular Formula: C26H21NO4S
InChi Key: BQIZNDWONIMCGM-HSZRJFAPSA-N
SMILES: OC(=O)[C@@H](CC1=CSC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)