accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17815-99-5

IUPAC Name: 4-chloro-N,N-dimethyl-2-nitroaniline
Synonyms: 4-chloro-N,N-dimethyl-2-nitroaniline
Molecular Weight (g/mol): 200.62
Molecular Formula: C8H9ClN2O2
InChi Key: QOMAYFFYZHAPEH-UHFFFAOYSA-N
SMILES: CN(C)C1=C(C=C(Cl)C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results