Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N,N-dimethyl-2-nitroaniline, 97%, Thermo Scientific™

Catalog No. AAH63169MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63169MD 250 mg
AAH6316903 1 g
AAH6316906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63169MD Supplier Thermo Scientific Chemicals Supplier No. H63169MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17815-99-5
Molecular Formula C8H9ClN2O2
Molecular Weight (g/mol) 200.622
InChI Key QOMAYFFYZHAPEH-UHFFFAOYSA-N
Synonym 4-chloro-2-nitro-phenyl-dimethyl-amine, 2-nitro-4-chloro-n,n-dimethylaniline, 4-chloro-n,n-dimethyl-2-nitrobenzenamine, benzenamine, 4-chloro-n,n-dimethyl-2-nitro
PubChem CID 12468660
IUPAC Name 4-chloro-N,N-dimethyl-2-nitroaniline
SMILES CN(C)C1=C(C=C(C=C1)Cl)[N+](=O)[O-]

Specifications

Quantity 250 mg
UN Number UN2811
Beilstein 2213297
Formula Weight 200.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-N,N-dimethyl-2-nitroaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.