accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 178162-69-1

IUPAC Name: 1-[(4-bromophenyl)methyl]piperidine
Synonyms: 1-[(4-bromophenyl)methyl]piperidine
Molecular Weight (g/mol): 254.17
Molecular Formula: C12H16BrN
InChi Key: ILQFFXGIIWWTNH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CN2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results