accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOOOHCl

CAS RN 1781835-20-8

IUPAC Name: 2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide hydrochloride
Synonyms: AdipoRon hydrochloride
Molecular Weight (g/mol): 464.99
Molecular Formula: C27H29ClN2O3
InChi Key: TZVJQEGKRLDTHQ-UHFFFAOYSA-N
SMILES: Cl.O=C(COC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)NC1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results