accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17829-86-6

IUPAC Name: samarium(3+) triacetate
Synonyms: samarium(3+) triacetate
Molecular Weight (g/mol): 327.49
Molecular Formula: C6H9O6Sm
InChi Key: JPDBEEUPLFWHAJ-UHFFFAOYSA-K
SMILES: [Sm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results