accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSH2N

CAS RN 1783-81-9

IUPAC Name: 3-(methylsulfanyl)aniline
Synonyms: M-methioaniline
Molecular Weight (g/mol): 139.22
Molecular Formula: C7H9NS
InChi Key: KCHLDNLIJVSRPK-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(N)=C1
Boiling Point 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)