Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)aniline 98.0+%, TCI America™
SDP

Catalog No. M167325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M167325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M167325G Supplier TCI America Supplier No. M167325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1783-81-9
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
InChI Key KCHLDNLIJVSRPK-UHFFFAOYSA-N
Synonym 3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
PubChem CID 15693
IUPAC Name 3-methylsulfanylaniline
SMILES CSC1=CC=CC(=C1)N

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
Formula Weight 139.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)aniline
RTECSNumber BY6301000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.