accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178305-99-2

IUPAC Name: {2-fluoro-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 2-fluoro-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 216.02
Molecular Formula: C12H10BFO2
InChi Key: BWYWXDFYJSIUBE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results