Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F07855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0785-5G 5 g
F07851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0785-5G Supplier TCI America Supplier No. F07855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 178305-99-2
Molecular Formula C12H10BFO2
MDL Number MFCD01075707
InChI Key BWYWXDFYJSIUBE-UHFFFAOYSA-N
PubChem CID 2774698

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 5 g
Synonym 2-fluoro-4-biphenylylboronic acid, 2-fluoro-4-biphenylboronic acid, 2-fluorobiphenyl-4-boronic acid, 2-fluorobiphenyl-4-ylboronic acid, 3-fluoro-4-phenylphenyl boranediol
SMILES OB(O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
IUPAC Name {2-fluoro-[1,1'-biphenyl]-4-yl}boronic acid
Molecular Weight (g/mol) 216.02
Formula Weight 216.02
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.