accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOCH2

CAS RN 17832-28-9

IUPAC Name: 4-(ethenyloxy)butan-1-ol
Synonyms: 4-(ethenyloxy)butan-1-ol
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: HMBNQNDUEFFFNZ-UHFFFAOYSA-N
SMILES: OCCCCOC=C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)