Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene Glycol Monovinyl Ether (stabilized with KOH) 97.0+%, TCI America™
SDP

Catalog No. T1796500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1796500ML 500 mL
T179625ML 25 mL
T1796100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1796500ML Supplier TCI America Supplier No. T1796500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17832-28-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00080697
InChI Key HMBNQNDUEFFFNZ-UHFFFAOYSA-N
Synonym 4-vinyloxy butan-1-ol, 1-butanol, 4-ethenyloxy, 4-hydroxybutyl vinyl ether, hydroxybutyl vinyl ether, unii-8lgm5rzx0n, 4-vinyloxybutanol, 4-ethenyloxy butan-1-ol, 8lgm5rzx0n, 1,4-butanediol monovinyl ether, hbuve
PubChem CID 87329
IUPAC Name 4-(ethenyloxy)butan-1-ol
SMILES OCCCCOC=C

Specifications

Color Yellow
Boiling Point 86°C
Quantity 500 mL
Formula Weight 116.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetramethylene Glycol Monovinyl Ether (stabilized with KOH)
EINECSNumber (2)-3248&(7)-0097
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.