accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CCH3OOHO

CAS RN 178324-24-8

IUPAC Name: tetrabutylazanium 2-hydroxypropanoate
Synonyms: tetrabutylammonium ion lactate
Molecular Weight (g/mol): 331.54
Molecular Formula: C19H41NO3
InChi Key: MSLTZKLJPHUCPU-UHFFFAOYNA-M
SMILES: CC(O)C([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results