Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium L-lactate, 70% w/w aq. soln., Thermo Scientific™

Catalog No. AA43035AC
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43035AC 25 mL
AA43035AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43035AC Supplier Thermo Scientific Chemicals Supplier No. 43035AC

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178324-24-8
Molecular Formula C19H41NO3
Molecular Weight (g/mol) 331.54
MDL Number MFCD00216638
InChI Key MSLTZKLJPHUCPU-UHFFFAOYNA-M
Synonym tetrabutylammonium lactate, tetra-n-butylammonium l-lactate, tetrabutylammonium ion lactate, tetrabutylammonium 2-hydroxypropanoate, tetrabutylammonium 2-hydroxy-propionate, 1-butanaminium, n,n,n-tributyl-, 2-hydroxypropanoate 1:1
PubChem CID 16218415
SMILES CC(O)C([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.957 g/mL
Quantity 25 mL
IUPAC Name (2S)-2-hydroxypropanoate;tetrabutylazanium
Formula Weight 331.54
Concentration or Composition (by Analyte or Components) 70% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-butylammonium L-lactate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.