accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1785-61-1

IUPAC Name: 1,3-diethynylbenzene
Synonyms: 1,3-diethynylbenzene
Molecular Weight (g/mol): 126.16
Molecular Formula: C10H6
InChi Key: PNXLPYYXCOXPBM-UHFFFAOYSA-N
SMILES: C#CC1=CC(=CC=C1)C#C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)