Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Diethynylbenzene

Catalog No. 6321045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-210-45G 5 g
63-210-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-210-45G Supplier Sigma Aldrich Supplier No. 6321045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Diethynylbenzene may be used to synthesize mono- and di- 1,4-substituted-1,2,3-triazole ligands, which have the property to self-assemble into metallo-cyclic structures in the presence of silver ions.

Specifications

CAS 1785-61-1
Density 0.949 g/mL (at 25°C (literature))
Boiling Point 188°C to 189°C (lit.)
Molecular Formula C10H6
Refractive Index n20/D 1.5820 (literature)
Linear Formula C10H6
MDL Number MFCD00041686
Quantity 5 g
Molecular Weight (g/mol) 126.15
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.