CAS RN 1785764-26-2
CAS RN 1785764-26-2
IUPAC Name:
5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride
Synonyms:
5,6-diamino-1,3-diethylpyrimidine-2,4-dione hydrochloride
Molecular Weight (g/mol):
234.68
Molecular Formula:
C8H15ClN4O2
InChi Key:
UQWLJOFXKBZWCI-UHFFFAOYSA-N
SMILES:
Cl.CCN1C(N)=C(N)C(=O)N(CC)C1=O
1
–
1
of
1
results
1
5,6-Diamino-1,3-diethyluracil Hydrochloride 98.0+%, TCI America™
CAS: 1785764-26-2 Molecular Formula: C8H15ClN4O2 Molecular Weight (g/mol): 234.68 MDL Number: MFCD22573561 InChI Key: UQWLJOFXKBZWCI-UHFFFAOYSA-N PubChem CID: 71299790 IUPAC Name: 5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride SMILES: Cl.CCN1C(N)=C(N)C(=O)N(CC)C1=O