accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17866-84-1

IUPAC Name: N-(4-fluorophenyl)-1-(4-{[(4-fluorophenyl)imino]methyl}phenyl)methanimine
Synonyms: N-(4-fluorophenyl)-1-(4-{[(4-fluorophenyl)imino]methyl}phenyl)methanimine
Molecular Weight (g/mol): 320.34
Molecular Formula: C20H14F2N2
InChi Key: VBCDABROGMPOGB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N=CC1=CC=C(C=NC2=CC=C(F)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results