Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthalbis(4-fluoroaniline) 98.0+%, TCI America™
SDP

Catalog No. B087510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B087510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B087510G Supplier TCI America Supplier No. B087510G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17866-84-1
Molecular Formula C20H14F2N2
Molecular Weight (g/mol) 320.343
MDL Number MFCD00017952
InChI Key VBCDABROGMPOGB-UHFFFAOYSA-N
Synonym Bis(4-fluoroanilino)terephthalaldehyde
PubChem CID 321189
IUPAC Name N-(4-fluorophenyl)-1-[4-[(4-fluorophenyl)iminomethyl]phenyl]methanimine
SMILES C1=CC(=CC=C1C=NC2=CC=C(C=C2)F)C=NC3=CC=C(C=C3)F

Specifications

Color Yellow
Quantity 10 g
Formula Weight 320.34
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalbis(4-fluoroaniline)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.