accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ONNCH3CH2CH3

CAS RN 178803-51-5

IUPAC Name: 4-{[2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](phenyl)methyl}-N,N-diethylbenzamide
Synonyms: 4-{[2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](phenyl)methyl}-N,N-diethylbenzamide
Molecular Weight (g/mol): 419.61
Molecular Formula: C27H37N3O
InChi Key: WGIDFDFAOQVAHY-UHFFFAOYNA-N
SMILES: CCN(CC)C(=O)C1=CC=C(C=C1)C(N1CC(C)N(CC=C)CC1C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results