accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17894-26-7

IUPAC Name: 2,5-dimethoxy-3-nitrobenzoate
Synonyms: 2,5-dimethoxy-3-nitrobenzoate
Molecular Weight (g/mol): 226.17
Molecular Formula: C9H8NO6
InChi Key: QCJROOYLFVYZEP-UHFFFAOYSA-M
SMILES: COC1=CC(C([O-])=O)=C(OC)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results