Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxy-3-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D29491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29491G Supplier TCI America Supplier No. D29491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17894-26-7
Molecular Formula C9H9NO6
Molecular Weight (g/mol) 227.172
MDL Number MFCD00017022
InChI Key QCJROOYLFVYZEP-UHFFFAOYSA-N
PubChem CID 2733991
SMILES COC1=CC(=C(C(=C1)C(=O)O)OC)[N+](=O)[O-]

Specifications

Melting Point 183°C
Color Yellow
Quantity 1 g
Formula Weight 227.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxy-3-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.