accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFOHOF

CAS RN 178974-97-5

IUPAC Name: 2,4-difluoro-3-methoxybenzoic acid
Synonyms: 2,4-difluoro-3-methoxybenzoic acid
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: KWLIGHXPUYUTBH-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(C(O)=O)=C1F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)