accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CH3CH2C

CAS RN 17898-21-4

IUPAC Name: triethyl(prop-2-en-1-yl)silane
Synonyms: triethyl(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 156.34
Molecular Formula: C9H20Si
InChi Key: SVGQCVJXVAMCPM-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results