Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltriethylsilane 97.0+%, TCI America™
SDP

Catalog No. A22995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22995G Supplier TCI America Supplier No. A22995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An organosilicon reagent used in organic synthesis.

Chemical Identifiers

CAS 17898-21-4
Molecular Formula C9H20Si
Molecular Weight (g/mol) 156.34
MDL Number MFCD18207713
InChI Key SVGQCVJXVAMCPM-UHFFFAOYSA-N
PubChem CID 11194462
IUPAC Name triethyl(prop-2-en-1-yl)silane
SMILES CC[Si](CC)(CC)CC=C

Specifications

Color Colorless
Boiling Point 168°C
Quantity 5 g
UN Number 1993
Formula Weight 156.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltriethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.