accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17920-85-3

IUPAC Name: [4-(furan-2-yl)phenyl]methanol
Synonyms: [4-(furan-2-yl)phenyl]methanol
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: IKZLKNNNCKXCKP-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results