accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONCO

CAS RN 1793-07-3

IUPAC Name: methyl 2-isocyanatobenzoate
Synonyms: methyl 2-isocyanatobenzoate
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: ATFKBWNEVZUSKC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results