accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 1795-15-9

IUPAC Name: octylcyclohexane
Synonyms: 1-cyclohexyloctane
Molecular Weight (g/mol): 196.38
Molecular Formula: C14H28
InChi Key: FBXWCEKQCVOOLT-UHFFFAOYSA-N
SMILES: CCCCCCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results