Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. O013825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O013825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O013825ML Supplier TCI America Supplier No. O013825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1795-15-9
Molecular Formula C14H28
Molecular Weight (g/mol) 196.378
MDL Number MFCD00039464
InChI Key FBXWCEKQCVOOLT-UHFFFAOYSA-N
Synonym 1-Cyclohexyloctane
PubChem CID 15712
IUPAC Name octylcyclohexane
SMILES CCCCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 264°C
Quantity 25 mL
Formula Weight 196.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.