accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 1795-16-0

IUPAC Name: decylcyclohexane
Synonyms: N-decylcyclohexane
Molecular Weight (g/mol): 224.43
Molecular Formula: C16H32
InChi Key: STWFZICHPLEOIC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results