Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. D123125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D123125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D123125ML Supplier TCI America Supplier No. D123125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1795-16-0
Molecular Formula C16H32
Molecular Weight (g/mol) 224.43
MDL Number MFCD00055277
InChI Key STWFZICHPLEOIC-UHFFFAOYSA-N
Synonym 1-Cyclohexyldecane
PubChem CID 15713
IUPAC Name decylcyclohexane
SMILES CCCCCCCCCCC1CCCCC1

Specifications

Melting Point -2°C
Color Colorless
Boiling Point 299°C
Quantity 25 mL
Formula Weight 224.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.