accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 1795-18-2

IUPAC Name: tetradecylcyclohexane
Synonyms: N-tetradecylcyclohexane
Molecular Weight (g/mol): 280.54
Molecular Formula: C20H40
InChi Key: NQAVPKIJZCHUNS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results